Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lhfpl3
Entrez Gene ID
100145306
Gene Name
LHFPL tetraspan subfamily member 3
Gene Aliases
LHFPL4
Chromosome Location
Chr.3: 67753381 - 67785612 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CX956539.2 - 1 - 2 219 106

View Full Product Details