Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tgm6
Entrez Gene ID
100145475
Gene Name
transglutaminase 6
Gene Aliases
-
Chromosome Location
Chr.10: 14814095 - 14828286 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126927.1 NP_001120399.1 1 - 2 208 106
GenBank mRNA BC161110.1 - 1 - 2 208 106

View Full Product Details