Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naa40
Entrez Gene ID
100145481
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
nat11
Chromosome Location
Chr.4: 40816569 - 40822316 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126933.1 NP_001120405.1 4 - 5 355 87
XM_012953533.2 XP_012808987.1 4 - 5 351 87
XM_012953535.2 XP_012808989.1 4 - 5 438 87
GenBank mRNA BC161120.1 - 4 - 5 355 87

View Full Product Details