Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clhc1
Entrez Gene ID
100145484
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
c2orf63
Chromosome Location
Chr.5: 20222297 - 20236576 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126935.1 NP_001120407.1 10 - 11 1480 104
XM_012962732.2 XP_012818186.1 10 - 11 1560 104
XM_012962733.2 XP_012818187.1 10 - 11 1626 104
XM_012962734.2 XP_012818188.1 10 - 11 1524 104
XM_012962735.2 XP_012818189.1 11 - 12 2031 104
XM_012962736.2 XP_012818190.1 11 - 12 2028 104
XM_012962737.1 XP_012818191.1 10 - 11 1517 104
XM_012962738.2 XP_012818192.2 8 - 9 1093 104
GenBank mRNA BC161123.1 - 10 - 11 1480 104

View Full Product Details