Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uts2b
Entrez Gene ID
100145570
Gene Name
urotensin 2B
Gene Aliases
uts2d
Chromosome Location
Chr.5: 99110499 - 99118722 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126995.1 NP_001120467.1 1 - 2 135 70
GenBank mRNA BC161294.1 - 1 - 2 135 70

View Full Product Details