Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pyroxd1
Entrez Gene ID
100145712
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016685229: 18 - 1328 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127086.1 NP_001120558.1 - 674 92
GenBank mRNA BC161508.1 - - 674 92

View Full Product Details