Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tcp11l2
Entrez Gene ID
100145732
Gene Name
t-complex 11, testis-specific-like 2
Gene Aliases
-
Chromosome Location
Chr.3: 38853423 - 38868455 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127106.1 NP_001120578.1 9 - 10 1513 92
GenBank mRNA BC161560.1 - 9 - 10 1513 92

View Full Product Details