Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mettl17
Entrez Gene ID
100145785
Gene Name
methyltransferase like 17
Gene Aliases
mett11d1
Chromosome Location
Chr.1: 130982400 - 130998793 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127148.1 NP_001120620.1 12 - 13 1142 115
GenBank mRNA BC161764.1 - 12 - 13 1142 115
CU075325.1 - 12 - 13 1146 115

View Full Product Details