Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
maats1
Entrez Gene ID
100151718
Gene Name
MYCBP associated and testis expressed 1
Gene Aliases
aat1, aat1alpha, c3orf15
Chromosome Location
Chr.Un|NW_016683629: 165623 - 189422 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127421.1 NP_001120893.1 12 - 13 1532 100
GenBank mRNA BC166178.1 - 12 - 13 1532 100

View Full Product Details