Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acsf3
Entrez Gene ID
100158470
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
Chromosome Location
Chr.4: 61046196 - 61067383 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127910.2 NP_001121382.2 8 - 9 1841 71
XM_012960641.2 XP_012816095.1 9 - 10 1972 71
XM_012960642.2 XP_012816096.1 9 - 10 2125 71
XM_012960643.2 XP_012816097.1 9 - 10 1977 71
XM_012960644.2 XP_012816098.1 8 - 9 1796 71
XM_012960645.2 XP_012816099.1 8 - 9 1870 71
XM_012960646.2 XP_012816100.1 8 - 9 1868 71
GenBank mRNA BC122927.1 - 8 - 9 1780 71
BC166077.1 - 9 - 10 1948 71
CT025405.2 - 8 - 9 1697 71

View Full Product Details