Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
retsat
Entrez Gene ID
100158500
Gene Name
retinol saturase (all-trans-retinol 13,14-reductase)
Gene Aliases
-
Chromosome Location
Chr.3: 107368085 - 107382564 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127940.1 NP_001121412.1 10 - 11 1700 67
XM_012958621.2 XP_012814075.1 11 - 12 1905 67
XM_012958622.1 XP_012814076.1 11 - 12 1904 67
GenBank mRNA BC166112.1 - 10 - 11 1700 67

View Full Product Details