Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nptx1
Entrez Gene ID
100158577
Gene Name
neuronal pentraxin 1
Gene Aliases
np1
Chromosome Location
Chr.10: 19107716 - 19114532 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128007.1 NP_001121479.1 4 - 5 1150 67
GenBank mRNA BC166253.1 - 4 - 5 1150 67

View Full Product Details