Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nop9
Entrez Gene ID
100158615
Gene Name
NOP9 nucleolar protein
Gene Aliases
c14orf21
Chromosome Location
Chr.8: 114701160 - 114709882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128031.1 NP_001121503.1 6 - 7 1146 93
GenBank mRNA BC166335.1 - 6 - 7 1146 93

View Full Product Details