Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nop9
Entrez Gene ID
100158615
Gene Name
NOP9 nucleolar protein
Gene Aliases
c14orf21
Chromosome Location
Chr.8: 114701160 - 114709882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128031.1 NP_001121503.1 9 - 10 1635 74
GenBank mRNA BC166335.1 - 9 - 10 1635 74

View Full Product Details