Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kank1
Entrez Gene ID
733504
Gene Name
KN motif and ankyrin repeat domains 1
Gene Aliases
ankrd15, ankrd15.2, kank1.2
Chromosome Location
Chr.1: 116510774 - 116628290 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128640.1 NP_001122112.1 12 - 13 4151 116
XM_012964011.2 XP_012819465.1 11 - 12 4376 116
XM_012964013.2 XP_012819467.1 11 - 12 4417 116
XM_012964015.2 XP_012819469.1 10 - 11 4155 116
XM_012964018.2 XP_012819472.1 10 - 11 4281 116
GenBank mRNA BC167678.1 - 12 - 13 4151 116

View Full Product Details