Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfap2e
Entrez Gene ID
100170172
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
ap2e
Chromosome Location
Chr.2: 56275511 - 56301612 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129928.1 NP_001123400.1 2 - 3 660 73
GenBank mRNA BC167362.1 - 2 - 3 660 73

View Full Product Details