Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ybx2
Entrez Gene ID
100170415
Gene Name
Y-box binding protein 2
Gene Aliases
contrin, csda, csda3, dbpc, frgy2, msy2, ybx2-a, ybx2-b
Chromosome Location
Chr.Un|NW_016684694: 5215 - 12547 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130195.1 NP_001123667.1 - 884 92
GenBank mRNA BC166950.1 - - 884 92

View Full Product Details