Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
c7orf57
Entrez Gene ID
100170423
Gene Name
chromosome 7 open reading frame 57
Gene Aliases
-
Chromosome Location
Chr.6: 68077505 - 68093607 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130201.1 NP_001123673.1 2 - 3 162 64
GenBank mRNA BC166964.1 - 2 - 3 162 64

View Full Product Details