Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
panx1
Entrez Gene ID
100170473
Gene Name
pannexin 1
Gene Aliases
mrs1, pannexin1, px1
Chromosome Location
Chr.2: 162365514 - 162389466 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130256.1 NP_001123728.1 4 - 5 1315 72
GenBank mRNA BC167372.1 - 4 - 5 1315 72

View Full Product Details