Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrk
Entrez Gene ID
100170575
Gene Name
SNF related kinase
Gene Aliases
-
Chromosome Location
Chr.6: 73142725 - 73187779 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130352.1 NP_001123824.1 5 - 6 1481 70
GenBank mRNA BC167632.1 - 5 - 6 1481 70
CT030415.1 - 5 - 6 1366 70

View Full Product Details