Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tfap4
Entrez Gene ID
100170601
Gene Name
transcription factor AP-4 (activating enhancer binding protein 4)
Gene Aliases
-
Chromosome Location
Chr.9: 14568711 - 14582394 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130369.1 NP_001123841.1 6 - 7 980 57
XM_012970169.2 XP_012825623.1 6 - 7 1542 57
XM_018097054.1 XP_017952543.1 5 - 6 1017 57
GenBank mRNA BC167689.1 - 6 - 7 980 57

View Full Product Details