Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccnj
Entrez Gene ID
100038087
Gene Name
cyclin J
Gene Aliases
ba690p14.1
Chromosome Location
Chr.7: 17641612 - 17672884 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134813.2 NP_001128285.1 4 - 5 738 78
XM_012965835.2 XP_012821289.1 4 - 5 1004 78
XM_012965836.2 XP_012821290.1 4 - 5 964 78
XM_012965837.2 XP_012821291.1 4 - 5 708 78
XM_012965838.2 XP_012821292.1 4 - 5 635 78
GenBank mRNA CT030404.1 - 4 - 5 720 78

View Full Product Details