Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrnp48
Entrez Gene ID
100216024
Gene Name
small nuclear ribonucleoprotein, U11/U12 48KDa subunit
Gene Aliases
-
Chromosome Location
Chr.6: 83249709 - 83258461 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142014.1 NP_001135486.1 3 - 4 337 101
GenBank mRNA BC167869.1 - 3 - 4 337 101

View Full Product Details