Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ficd
Entrez Gene ID
100216031
Gene Name
FIC domain containing
Gene Aliases
hype, unq3041
Chromosome Location
Chr.1: 133813018 - 133818600 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142020.1 NP_001135492.1 2 - 3 377 97
XM_012957346.2 XP_012812800.1 2 - 3 469 97
XM_012957351.2 XP_012812805.1 2 - 3 441 97
XM_018091319.1 XP_017946808.1 2 - 3 1202 97
GenBank mRNA BC167892.1 - 2 - 3 377 97

View Full Product Details