Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abhd2
Entrez Gene ID
100216078
Gene Name
abhydrolase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 99654934 - 99691049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142063.1 NP_001135535.1 9 - 10 1114 85
XM_018091965.1 XP_017947454.1 8 - 9 1077 85
GenBank mRNA BC168004.1 - 9 - 10 1114 85

View Full Product Details