Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnf8
Entrez Gene ID
100216121
Gene Name
ring finger protein 8, E3 ubiquitin protein ligase
Gene Aliases
rnf8-a, rnf8-b
Chromosome Location
Chr.Un|NW_016683871: 7733 - 17984 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142100.2 NP_001135572.1 7 - 8 665 74
XM_012954789.2 XP_012810243.1 6 - 7 1672 74
XM_012954790.2 XP_012810244.1 6 - 7 1672 74
GenBank mRNA BC168076.1 - 7 - 8 667 74

View Full Product Details