Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxo45
Entrez Gene ID
100216147
Gene Name
F-box protein 45
Gene Aliases
-
Chromosome Location
Chr.5: 126207941 - 126216356 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142120.1 NP_001135592.1 3 - 4 867 85
GenBank mRNA BC168120.1 - 3 - 4 867 85

View Full Product Details