Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vash1
Entrez Gene ID
100216209
Gene Name
vasohibin 1
Gene Aliases
-
Chromosome Location
Chr.8: 61359303 - 61372225 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142178.1 NP_001135650.1 6 - 7 1230 80
XM_012968117.2 XP_012823571.1 4 - 5 1614 80
GenBank mRNA BC168537.1 - 6 - 7 1230 80

View Full Product Details