Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a16
Entrez Gene ID
100216243
Gene Name
solute carrier family 25 member 16
Gene Aliases
d10s105e, gda, gdc, hgt.1, hml7, ml7
Chromosome Location
Chr.7: 30450397 - 30470170 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142210.1 NP_001135682.1 4 - 5 413 92
XM_012966233.2 XP_012821687.1 4 - 5 743 92
XM_012966234.2 XP_012821688.1 4 - 5 621 92
XM_012966235.2 XP_012821689.1 4 - 5 626 92
GenBank mRNA BC168609.1 - 4 - 5 413 92

View Full Product Details