Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sfxn3
Entrez Gene ID
100216274
Gene Name
sideroflexin 3
Gene Aliases
-
Chromosome Location
Chr.7: 21568263 - 21586416 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142227.1 NP_001135699.1 10 - 11 1366 97
XM_012966244.2 XP_012821698.1 10 - 11 988 97
GenBank mRNA BC168812.1 - 10 - 11 1366 97

View Full Product Details