Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc50
Entrez Gene ID
100216280
Gene Name
unc-50 homolog
Gene Aliases
gmh1, unc50-a, unc50-b, uncl, urp
Chromosome Location
Chr.2: 109445656 - 109456060 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142233.1 NP_001135705.1 4 - 5 568 110
XM_012956580.2 XP_012812034.1 5 - 6 746 110
XM_012956581.2 XP_012812035.1 4 - 5 689 110
GenBank mRNA BC168819.1 - 4 - 5 568 110
BC169164.1 - 4 - 5 568 110

View Full Product Details