Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
epgn
Entrez Gene ID
100216288
Gene Name
epithelial mitogen
Gene Aliases
epg, epigen
Chromosome Location
Chr.Un|NW_016683393: 83556 - 88981 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142239.2 NP_001135711.2 2 - 3 138 121
GenBank mRNA BC168838.1 - 2 - 3 132 121
BX703151.1 - 2 - 3 137 121
CU025080.1 - 2 - 3 138 121
DR879072.1 - 2 - 3 114 121

View Full Product Details