Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
henmt1
Entrez Gene ID
100286818
Gene Name
HEN methyltransferase 1
Gene Aliases
c1orf59, hen1, pimet1
Chromosome Location
Chr.4: 102214480 - 102229934 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159540.1 NP_001153012.1 1 - 2 90 102
XM_012960696.2 XP_012816150.1 2 - 3 421 102
XM_012960697.2 XP_012816151.1 2 - 3 275 102
XM_012960698.2 XP_012816152.1 2 - 3 207 102
GenBank mRNA EU338244.1 - 1 - 2 90 102

View Full Product Details