Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdc42
Entrez Gene ID
493389
Gene Name
cell division cycle 42
Gene Aliases
Xcdc42, cdc42hs, g25k
Chromosome Location
Chr.7: 68578504 - 68619101 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017070.4 NP_001017070.1 - 2447 98
GenBank mRNA BC122003.1 - 5 - 5 1144 98
CR761548.2 - 5 - 5 1117 98
CT030052.3 - 5 - 5 1180 98

View Full Product Details