Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bloc1s1
Entrez Gene ID
779698
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
Gene Aliases
blos1, gcn5l1
Chromosome Location
Chr.2: 133812251 - 133827344 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017271.2 NP_001017271.1 3 - 3 446 96
GenBank mRNA CR855794.2 - 3 - 3 446 96

View Full Product Details