Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cacng6
Entrez Gene ID
100125205
Gene Name
calcium channel, voltage-dependent, gamma subunit 6
Gene Aliases
-
Chromosome Location
Chr.7: 108448784 - 108456282 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103204.1 NP_001096674.1 - 448 100
GenBank mRNA BC136102.1 - - 448 100

View Full Product Details