Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrn1
Entrez Gene ID
100145075
Gene Name
neuritin 1
Gene Aliases
cpg15, neuritin, nrn, nrn1-a, nrn1-b
Chromosome Location
Chr.6: 49842702 - 49851398 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126599.1 NP_001120071.1 2 - 2 353 88
GenBank mRNA BC158490.1 - 2 - 2 353 88

View Full Product Details