The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203580.1 | NP_988911.1 | 2 - 2 | 650 | 64 |
GenBank mRNA | BC059770.1 | - | 2 - 2 | 650 | 64 |
CR760778.2 | - | 2 - 2 | 620 | 64 |