Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Atg12
Entrez Gene ID
100240693
Gene Name
autophagy-related 12
Gene Aliases
ZEAMMB73_234520
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144019.1 NP_001137491.1 - 24 54
GenBank mRNA BT063792.1 - - 291 54
FJ445015.1 - - 24 54
FJ445016.1 - - 24 54

View Full Product Details