Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100281664
Entrez Gene ID
100281664
Gene Name
ATBRCA1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001154584.1 NP_001148056.1 - 1098 71
GenBank mRNA EU957485.1 - - 1098 71

View Full Product Details