Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sfp1
Entrez Gene ID
541917
Gene Name
sulfate permease1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111580.1 NP_001105050.1 - 1181 82
GenBank mRNA AF355602.1 - - 1181 82

View Full Product Details