Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cesa10
Entrez Gene ID
542685
Gene Name
cellulose synthase10
Gene Aliases
ZmCESA10
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112202.1 NP_001105672.1 - 2323 75
GenBank mRNA AY372244.1 - - 2323 75

View Full Product Details