Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
psy2
Entrez Gene ID
542686
Gene Name
phytoene synthase2
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114645.1 NP_001108117.1 - 635 59
GenBank mRNA AY450646.1 - - 612 59
AY773476.1 - - 635 59
BT039824.1 - - 796 59
BT053947.1 - - 714 59

View Full Product Details