Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100191272
Entrez Gene ID
100191272
Gene Name
4-nitrophenylphosphatase
Gene Aliases
ZEAMMB73_060040
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136706.1 NP_001130178.1 - 617 115
GenBank mRNA BT033318.1 - - 617 115

View Full Product Details