Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
dhs2
Entrez Gene ID
100191910
Gene Name
deoxyhypusine synthase 2
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137334.1 NP_001130806.1 - 501 62
GenBank mRNA BT034162.1 - - 501 62
GU735678.1 - - 403 62

View Full Product Details