Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tan1
Entrez Gene ID
542642
Gene Name
tangled1
Gene Aliases
ZEAMMB73_420432
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112166.1 NP_001105636.1 - 184 96
GenBank mRNA AF305892.1 - - 184 96
BT069123.1 - - 109 96

View Full Product Details