Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lac5
Entrez Gene ID
732847
Gene Name
laccase-like
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112451.1 NP_001105921.1 - 593 81
GenBank mRNA AM086217.1 - - 593 81

View Full Product Details