Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PK
Entrez Gene ID
542177
Gene Name
PK protein
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111796.1 NP_001105266.1 - 393 71
GenBank mRNA AY623894.1 - - 393 71

View Full Product Details