Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pep4
Entrez Gene ID
542393
Gene Name
phosphoenolpyruvate carboxylase4
Gene Aliases
pep
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111968.1 NP_001105438.1 - 1079 82
GenBank mRNA X61489.1 - - 1079 82

View Full Product Details