Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rps11
Entrez Gene ID
542552
Gene Name
ribosomal protein S11
Gene Aliases
S11, ZEAMMB73_050662, ZmS11, cytosolic
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112092.1 NP_001105562.1 - 123 78
GenBank mRNA BT065528.1 - - 123 78
BT065599.1 - - 156 78
EU959120.1 - - 131 78
X55967.1 - - 123 78

View Full Product Details